Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoro-α-methylbenzyl alcohol 97%

Supplier: AMBEED, INC
Warning

A565781-5G A565781-100G A565781-10G A565781-1G A565781-5G
103839-910EA 9 USD InStock
103839-910 77197-690 77197-692 77197-694 77307-826
4-Fluoro-α-methylbenzyl alcohol 97%
4-Fluoro-α-methylbenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₉FO
Boiling Pt: 216.2 °C (760 mm Hg)
Melting Pt: 9.3 °C at N/A mmHg
Flash Pt: 90.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004515
CAS Number: 403-41-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty