Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(5-Fluoropyridin-3-yl)ethanone 96%

Supplier: AMBEED, INC
Warning

A119588-1G A119588-250MG A119588-5G
103799-788EA 118 USD InStock
103799-788 76983-156 77297-696
1-(5-Fluoropyridin-3-yl)ethanone 96%
1-(5-Fluoropyridin-3-yl)ethanone
Precaución: for Research Use Only


Formula: C₇H₆FNO
Boiling Pt: 213.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07375075
CAS Number: 342602-55-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty