Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Aminomethyl)cyclopropanol 95%

Supplier: AMBEED, INC
Warning

A139866-1G A139866-100MG A139866-250MG A139866-25G
103802-786EA 33 USD InStock
103802-786 77190-348 77190-350 77190-352 77190-354
1-(Aminomethyl)cyclopropanol 95%
1-(Aminomethyl)cyclopropanol
Precaución: for Research Use Only


Formula: C₄H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD04972522
CAS Number: 74592-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty