Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyloxy-3-bromobenzene 98%

Supplier: AMBEED, INC
Warning

A200383-25G A200383-5G A200383-100G A200383-10G A200383-500G
103847-824EA 5 USD InStock
103847-824 77085-882 77085-884 77274-794 77274-796 77274-798
1-Benzyloxy-3-bromobenzene 98%
1-Benzyloxy-3-bromobenzene
Precaución: for Research Use Only


Formula: C₁₃H₁₁BrO
MW: 263.13
Boiling Pt: 333.5 °C (760 mm Hg)
Melting Pt: 57…60 °C
Flash Pt: 137.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00155065
CAS Number: 53087-13-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty