Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A129828-1G A129828-100G A129828-10G A129828-250MG A129828-1G A129828-25G A129828-5G
103801-336EA 9 USD InStock
103801-336 76767-150 76767-152 76767-154 77313-724 77313-726 77313-728
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid 98%
1-(Benzyloxycarbonylamino)cyclopropyl-1-carboxylic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD00083285
CAS Number: 84677-06-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty