Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3-nitrobenzyl chloride 97%

Supplier: AMBEED, INC
Danger

A269445-1G A269445-5G A269445-100MG A269445-10G A269445-1G
103812-964EA 57 USD InStock
103812-964 103852-870 76827-290 76827-292 77092-636
2-Fluoro-3-nitrobenzyl chloride 97%
2-Fluoro-3-nitrobenzyl chloride
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09955448
CAS Number: 1020718-00-6
UN: 3261
ADR: 8,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty