Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Ethoxycarbonylcyclopropanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A103641-1G A103641-5G A103641-25G A103641-5G A103641-1G A103641-250MG
103796-592EA 9 USD InStock
103796-592 103822-300 77188-676 77283-138 77309-170 77745-789
1-Ethoxycarbonylcyclopropanecarboxylic acid 98%
1-Ethoxycarbonylcyclopropanecarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₁₀O₄
Boiling Pt: 259.1±23.0 °C (760 mm Hg)
Flash Pt: 105.8 °C
Storage Temperature: Ambient
MDL Number: MFCD08690114
CAS Number: 3697-66-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty