Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2'-Methylacetophenone 98%

Supplier: AMBEED, INC
Warning

A214174-1000G A214174-100G A214174-10G A214174-500G A214174-25G
103809-496EA 139 USD InStock
103809-496 77087-586 77112-524 77112-526 77112-528 77303-328
2'-Methylacetophenone 98%
2'-Methylacetophenone
Precaución: for Research Use Only


Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 214
Flash Pt: 75.55 °C
Storage Temperature: Ambient
MDL Number: MFCD00008734
CAS Number: 577-16-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty