Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A181932-1G A181932-5G A181932-1G A181932-100MG A181932-250MG A181932-5G
103807-668EA 83 USD InStock
103807-668 103846-038 77083-564 77151-632 77151-634 77301-760
1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester 95%
1-(3-Oxetanyl)-1H-pyrazole-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₉BN₂O₃
Boiling Pt: 384.7±32.0 °C at N/A mmHg
Flash Pt: 186.4 °C
Storage Temperature: Freezer
MDL Number: MFCD21337762
CAS Number: 1339890-99-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty