Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(4-Methylphenyl)cyclopentanecarboxylic acid 95%

Supplier: AMBEED, INC
Warning

A705876-5G A705876-25G
103840-664EA 65 USD InStock
103840-664 76844-874 77308-170
1-(4-Methylphenyl)cyclopentanecarboxylic acid 95%
1-(4-Methylphenyl)cyclopentanecarboxylic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₆O₂
Boiling Pt: 355.7 °C (760 mm Hg)
Melting Pt: 181…184 °C (760 mm Hg)
Flash Pt: 165.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00044672
CAS Number: 80789-75-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty