Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(Pyridin-2-yl)piperidin-4-amine 98%

Supplier: AMBEED, INC
Warning

A152355-1G A152355-5G A152355-10G A152355-250MG A152355-1G A152355-5G
103804-378EA 150 USD InStock
103804-378 103842-546 77235-474 77235-476 77318-158 77318-160
1-(Pyridin-2-yl)piperidin-4-amine 98%
1-(Pyridin-2-yl)piperidin-4-amine
Precaución: for Research Use Only


Formula: C₁₀H₁₅N₃
Boiling Pt: 318.7 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD08061082
CAS Number: 144465-94-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty