Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A167647-1G A167647-10G A167647-250MG A167647-1G A167647-25G A167647-5G
103806-204EA 12 USD InStock
103806-204 103844-462 77011-486 77011-488 77081-482 77081-484 77081-486
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid 95%
1-(tert-Butoxycarbonyl)azetidine-2-carboxylic acid
Precaución: for Research Use Only


Formula: C₉H₁₅NO₄
MW: 201.2197 g/mol
Boiling Pt: 321 °C (760 mm Hg)
Flash Pt: 147.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01861756
CAS Number: 159749-28-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty