Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyl-4-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A150077-1G A150077-5G A150077-100G A150077-10G A150077-500G A150077-1G A150077-25G A150077-5G
103804-118EA 6 USD InStock
103804-118 103842-252 77235-408 77235-410 77235-412 77317-760 77317-762 77317-764
1-tert-Butyl-4-iodobenzene 98%
1-tert-Butyl-4-iodobenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₃I
MW: 260.12
Boiling Pt: 114
Density: 1.4392
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00052339
CAS Number: 35779-04-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty