Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene 97%

Supplier: AMBEED, INC
Warning

A226627-1G A226627-5G A226627-25G A226627-10G A226627-1G
103819-254EA 8 USD InStock
103819-254 103850-158 77088-892 77193-016 77303-772
1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene 97%
1,2,3,4-Tetrahydro-1,1,4,4,6-pentamethylnaphthalene
Precaución: for Research Use Only


Formula: C₁₅H₂₂
Boiling Pt: 267.4±20.0 °C (760 mm Hg)
Melting Pt: 31 °C
Flash Pt: 112.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00833394
CAS Number: 6683-48-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty