Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,10-Dichlorodecane 97%

Supplier: AMBEED, INC
Danger

A107164-1000G A107164-100G A107164-10G
103807-964EA 141 USD InStock
103807-964 103822-904 77188-850 77188-852 77188-854
1,10-Dichlorodecane 97%
1,10-Dichlorodecane
Precaución: for Research Use Only


Formula: ClH₂C(CH₂)₈CH₂Cl
Boiling Pt: 284.3 °C (760 mm Hg)
Melting Pt: 15.6 °C
Flash Pt: 108.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00000957
CAS Number: 2162-98-3
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty