Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Diacetylferrocene 97%

Supplier: AMBEED, INC
Warning

A137380-1G A137380-100G A137380-10G A137380-1G A137380-5G
103802-450EA 6 USD InStock
103802-450 77190-248 77190-250 77298-906 77298-908 77315-406
1,1'-Diacetylferrocene 97%
1,1'-Diacetylferrocene
Precaución: for Research Use Only


Formula: C₁₄H₆FeO₂
Storage Temperature: Ambient
MDL Number: MFCD00001424
CAS Number: 1273-94-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty