Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Thiocarbonyldi-2(1H)-pyridone 95%

Supplier: AMBEED, INC
Warning

A141313-1G A141313-100G A141313-10G A141313-1G A141313-25G A141313-5G
103802-976EA 5 USD InStock
103802-976 77190-440 77190-442 77316-120 77316-122 77316-124
1,1'-Thiocarbonyldi-2(1H)-pyridone 95%
1,1'-Thiocarbonyldi-2(1H)-pyridone


Formula: C₁₁H₈N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD00075238
CAS Number: 102368-13-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty