Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pentamethylbenzene 98%

Supplier: AMBEED, INC
Danger

A777498-100G A777498-10G A777498-500G A777498-25G A777498-5G A777498-1G
103840-960EA 12 USD InStock
103840-960 76728-312 76728-314 76728-316 76845-220 76845-222 77308-310
Pentamethylbenzene 98%
Pentamethylbenzene
Precaución: for Research Use Only


Formula: C₁₁H₁₆
MW: 148.25 g/mol
Boiling Pt: 232
Melting Pt: 49…51 °C
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008522
CAS Number: 700-12-9
UN: 1325
ADR: 4.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty