Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3,4-Tetrahydroisoquinoline-6-carbonitrile hydrochloride 98%

Supplier: AMBEED, INC
Warning

A353671-1G A353671-5G A353671-100MG A353671-10G A353671-250MG A353671-1G
103826-104EA 99 USD InStock
103826-104 103837-106 77195-224 77195-226 77195-228 77306-650
1,2,3,4-Tetrahydroisoquinoline-6-carbonitrile hydrochloride 98%
1,2,3,4-Tetrahydroisoquinoline-6-carbonitrile hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₁ClN₂
Boiling Pt: 332.8 °C (760 mm Hg)
Melting Pt: 292…296 °C
Storage Temperature: Ambient
MDL Number: MFCD08234661
CAS Number: 171084-93-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty