Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2,3,4-Tetrahydroquinoline-6-carbonitrile 97%

Supplier: AMBEED, INC
Warning

A278123-1G A278123-5G A278123-100MG A278123-250MG A278123-1G
103813-392EA 117 USD InStock
103813-392 103853-330 76988-438 76988-440 77305-290
1,2,3,4-Tetrahydroquinoline-6-carbonitrile 97%
1,2,3,4-Tetrahydroquinoline-6-carbonitrile
Precaución: for Research Use Only


Formula: C₁₀H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD14534464
CAS Number: 50741-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty