Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diaminofurazan 97%

Supplier: AMBEED, INC
Warning

A188440-1G A188440-1G A188440-25G A188440-5G A188440-100G A188440-10G A188440-250MG
103816-098EA 6 USD InStock
103816-098 103846-694 77084-430 77084-432 77084-434 77274-360 77274-362 77274-364
3,4-Diaminofurazan 97%
3,4-Diaminofurazan
Precaución: for Research Use Only


Formula: C₂H₄N₄O
MW: 100.08 g/mol
Boiling Pt: 304 °C (760 mm Hg)
Melting Pt: 178…183 °C
Flash Pt: 137.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00138084
CAS Number: 17220-38-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty