Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2'-Dinitrodibenzyl 97%

Supplier: AMBEED, INC
Warning

A104004-1000G A104004-100G A104004-10G A104004-25G A104004-500G A104004-5G
103807-886EA 275 USD InStock
103807-886 77245-290 77245-292 77245-294 77245-296 77245-298
2,2'-Dinitrodibenzyl 97%
2,2'-Dinitrodibenzyl
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂O₄
Boiling Pt: 391.6 °C (760 mm Hg)
Melting Pt: 122 °C
Flash Pt: 178.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00024296
CAS Number: 16968-19-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty