Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Di(pyridin-4-yl)ethyne 95%

Supplier: AMBEED, INC
Warning

A174611-1G A174611-5G A174611-100MG A174611-1G A174611-5G A174611-250MG
103806-960EA 90 USD InStock
103806-960 103845-260 76974-194 77301-346 77301-348 77593-092
1,2-Di(pyridin-4-yl)ethyne 95%
1,2-Di(pyridin-4-yl)ethyne
Precaución: for Research Use Only


Formula: C₁₂H₈N₂
Boiling Pt: 350.9 °C (760 mm Hg)
Flash Pt: 155.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD17013087
CAS Number: 73564-69-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty