Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Dichloro-3-iodobenzene 98%

Supplier: AMBEED, INC
Warning

A756525-25G A756525-5G A756525-100G A756525-10G A756525-1G
103840-872EA 6 USD InStock
103840-872 76845-102 76845-104 77105-292 77105-294 77105-296
1,2-Dichloro-3-iodobenzene 98%
1,2-Dichloro-3-iodobenzene
Precaución: for Research Use Only


Formula: C₆H₃Cl₂I
MW: 272.9 g/mol
Boiling Pt: 242
Melting Pt: 35
Flash Pt: 114.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001034
CAS Number: 2401-21-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty