Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Veratrole 97%

Supplier: AMBEED, INC
Warning

A232020-1000G A232020-100G A232020-500G A232020-25G A232020-1000G
103809-638EA 61 USD InStock
103809-638 76786-292 76786-294 77089-484 77303-926
Veratrole 97%
Veratrole
Precaución: for Research Use Only


Formula: C₈H₁₀O₂
Boiling Pt: 206.3 °C (760 mm Hg)
Melting Pt: 15 °C
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00008357
CAS Number: 91-16-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty