Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,2-Oxazinane hydrochloride 97%

Supplier: AMBEED, INC
Danger

A147022-1G A147022-5G A147022-100MG A147022-250MG A147022-1G
103803-726EA 59 USD InStock
103803-726 103841-822 76997-150 76997-152 77299-556
1,2-Oxazinane hydrochloride 97%
1,2-Oxazinane hydrochloride
Precaución: for Research Use Only


Formula: C₄H₁₀ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD13193208
CAS Number: 54722-74-6
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty