Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Benzenediboronic acid bis(pinacol) ester 97%

Supplier: AMBEED, INC
Warning

A153212-1G A153212-5G A153212-10G A153212-250MG A153212-1G A153212-25G A153212-5G
103804-476EA 6 USD InStock
103804-476 103842-646 76812-332 76812-334 77318-282 77318-284 77318-286
1,3-Benzenediboronic acid bis(pinacol) ester 97%
1,3-Benzenediboronic acid bis(pinacol) ester
Precaución: for Research Use Only


Formula: C₁₈H₂₈B₂O₄
Boiling Pt: 430.9 °C (760 mm Hg)
Flash Pt: 214.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09864185
CAS Number: 196212-27-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty