Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diaminoguanidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A177212-5G A177212-100G A177212-10G A177212-25G A177212-500G
103845-544EA 5 USD InStock
103845-544 76974-286 76974-288 77082-930 77082-932 77881-348
1,3-Diaminoguanidine hydrochloride 98%
1,3-Diaminoguanidine hydrochloride
Precaución: for Research Use Only


Formula: CH₇N₅
Flash Pt: 86.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00012948
CAS Number: 36062-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty