Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Dibromoanisole 98%

Supplier: AMBEED, INC
Warning

A141477-1G A141477-10G A141477-1G A141477-25G A141477-5G
103803-002EA 13 USD InStock
103803-002 77246-972 77316-166 77316-168 77316-170
2,6-Dibromoanisole 98%
2,6-Dibromoanisole
Precaución: for Research Use Only


Formula: C₇H₆Br₂O
Boiling Pt: 253.7 °C (760 mm Hg)
Melting Pt: 13 °C
Flash Pt: 99.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00093270
CAS Number: 38603-09-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty