Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dibromo-5-tert-butylbenzene 95%

Supplier: AMBEED, INC
Warning

A226783-1G A226783-5G A226783-100G A226783-10G A226783-1G A226783-5G A226783-25G
103819-268EA 13 USD InStock
103819-268 103850-176 76786-134 76786-136 77088-926 77088-928 77303-778
1,3-Dibromo-5-tert-butylbenzene 95%
1,3-Dibromo-5-tert-butylbenzene
Precaución: for Research Use Only


Formula: C₁₀H₁₂Br₂
Boiling Pt: 250 °C (760 mm Hg)
Flash Pt: 115.6 °C
Storage Temperature: Ambient
MDL Number: MFCD06657956
CAS Number: 129316-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty