Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diethylurea 97%

Supplier: AMBEED, INC
Warning

A146337-5G A146337-100G A146337-10G A146337-5G A146337-25G
103841-742EA 7 USD InStock
103841-742 77190-632 77190-634 77299-512 77317-068
1,3-Diethylurea 97%
1,3-Diethylurea
Precaución: for Research Use Only


Formula: C₅H₁₂N₂O
Boiling Pt: 263 °C (760 mm Hg)
Melting Pt: 112…113 °C
Flash Pt: 121.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00009028
CAS Number: 623-76-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty