Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diiodobenzene 97%

Supplier: AMBEED, INC
Warning

A127837-1000G A127837-100G A127837-10G A127837-500G A127837-25G A127837-5G
77109-020EA 641.69 USD InStock
77109-020 77109-022 77109-024 77109-026 77313-296 77313-298
1,3-Diiodobenzene 97%
1,3-Diiodobenzene
Precaución: for Research Use Only


Formula: C₆H₄I₂
Boiling Pt: 285.7 °C (760 mm Hg)
Melting Pt: 34…37 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041731
CAS Number: 626-00-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty