Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Diisopropoxybenzene 98%

Supplier: AMBEED, INC
Warning

A182490-1G A182490-5G A182490-100G A182490-1G A182490-25G A182490-5G
103807-720EA 10 USD InStock
103807-720 103846-092 77138-774 77301-792 77301-794 77301-796
1,3-Diisopropoxybenzene 98%
1,3-Diisopropoxybenzene
Precaución: for Research Use Only


Formula: C₁₂H₁₈O₂
Boiling Pt: 89.5…95 °C (3 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784412
CAS Number: 79128-08-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty