Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethyl-1H-pyrazol-5-ol 97%

Supplier: AMBEED, INC
Warning

A343755-1G A343755-5G A343755-25G
103825-892EA 5 USD InStock
103825-892 103836-870 76839-912 76839-914 77306-562
1,3-Dimethyl-1H-pyrazol-5-ol 97%
1,3-Dimethyl-1H-pyrazol-5-ol
Precaución: for Research Use Only


Formula: C₅H₈N₂O
MW: 112.1300 g/mol
Boiling Pt: 225.9 °C (760 mm Hg)
Melting Pt: 113…117 °C
Flash Pt: 90 °C
Storage Temperature: Ambient
MDL Number: MFCD00129673
CAS Number: 5203-77-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty