Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Dimethyluracil-5-carboxaldehyde 96%

Supplier: AMBEED, INC
Warning

A161703-1G A161703-5G A161703-100MG A161703-250MG A161703-1G
103805-526EA 125 USD InStock
103805-526 103843-768 77191-096 77191-098 77319-768
1,3-Dimethyluracil-5-carboxaldehyde 96%
1,3-Dimethyluracil-5-carboxaldehyde
Precaución: Inert atmosphere, 2…8 °C.


Formula: C₇H₈N₂O₃
Boiling Pt: 274.1 °C (760 mm Hg)
Flash Pt: 122.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00099582
CAS Number: 4869-46-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty