Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dichloropyrido[3,4-d]pyridazine 97%

Supplier: AMBEED, INC
Warning

A601042-5G A601042-1G A601042-100MG A601042-250MG A601042-5G
103828-978EA 37 USD InStock
103828-978 103840-116 76844-278 77198-008 77198-010 77307-918
1,4-Dichloropyrido[3,4-d]pyridazine 97%
1,4-Dichloropyrido[3,4-d]pyridazine
Precaución: for Research Use Only


Formula: C₇H₃Cl₂N₃
Boiling Pt: 440.7 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09839782
CAS Number: 14490-19-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty