Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dimethylpyrazole 95%

Supplier: AMBEED, INC
Warning

A119305-1G A119305-100G A119305-25G A119305-1G
103799-750EA 9 USD InStock
103799-750 77189-560 77189-562 77297-674
1,4-Dimethylpyrazole 95%
1,4-Dimethylpyrazole
Precaución: for Research Use Only


Formula: C₅H₈N₂
Boiling Pt: 145.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01075158
CAS Number: 1072-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty