Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,5-Dibromo-2,3,4-trifluorobenzene 95%

Supplier: AMBEED, INC
Warning

A202536-1G A202536-5G A202536-100MG A202536-250MG A202536-25G A202536-5G
103817-398EA 31 USD InStock
103817-398 103848-042 77012-704 77012-706 77012-708 77086-152
1,5-Dibromo-2,3,4-trifluorobenzene 95%
1,5-Dibromo-2,3,4-trifluorobenzene
Precaución: for Research Use Only


Formula: C₆HBr₂F₃
MW: 289.875 g/mol
Boiling Pt: 200.9±35.0 °C (760 mm Hg)
Flash Pt: 75.3 °C
Storage Temperature: Ambient
MDL Number: MFCD28505052
CAS Number: 17299-95-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty