Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibenzylideneacetone 98%

Supplier: AMBEED, INC
Warning

A151381-1000G A151381-100G A151381-10G A151381-500G A151381-1000G A151381-25G
103808-756EA 287 USD InStock
103808-756 77028-136 77028-138 77028-140 77028-142 77299-872 77317-962
Dibenzylideneacetone 98%
Dibenzylideneacetone
Precaución: for Research Use Only


Formula: C₁₇H₁₄O
MW: 234.2900 g/mol
Boiling Pt: 400.7 °C (760 mm Hg)
Melting Pt: 107…113 °C
Flash Pt: 176.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004790
CAS Number: 538-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty