Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

12-Bromo-1-dodecanol 97%

Supplier: AMBEED, INC
Warning

A138140-1G A138140-100G A138140-10G A138140-1G A138140-25G A138140-5G
103802-562EA 5 USD InStock
103802-562 76972-444 76972-446 76972-448 77298-962 77315-582 77315-584
12-Bromo-1-dodecanol 97%
12-Bromo-1-dodecanol
Precaución: for Research Use Only


Formula: HO(CH₂)₁₁CH₂Br
Boiling Pt: 155 °C (760 mm Hg)
Melting Pt: 34…36 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004754
CAS Number: 3344-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty