Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

14-Hydroxytetradecanoic acid 95%

Supplier: AMBEED, INC
Warning

A209054-1G A209054-250MG A209054-50MG A209054-100MG
103817-908EA 112 USD InStock
103817-908 76975-472 76975-474 77671-852
14-Hydroxytetradecanoic acid 95%
14-Hydroxytetradecanoic acid
Precaución: for Research Use Only


Formula: C₁₄H₂₈O₃
MW: 244.37 g/mol
Boiling Pt: 387.1 °C (760 mm Hg)
Flash Pt: 202.1 °C
Storage Temperature: Ambient
MDL Number: MFCD19707605
CAS Number: 17278-74-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty