Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

16-Bromopalmitic acid 97%

Supplier: AMBEED, INC
Warning

A237188-1G A237188-5G A237188-1G A237188-5G A237188-250MG A237188-25G
103811-136EA 22 USD InStock
103811-136 103850-914 77089-986 77089-988 77275-932 77275-934
16-Bromopalmitic acid 97%
16-Bromopalmitic acid
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₁₄COOH
Boiling Pt: 422.9 °C (760 mm Hg)
Melting Pt: 68…71 °C
Flash Pt: 209.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00040779
CAS Number: 2536-35-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty