Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Azaspiro[4.5]decane-2,8-dione 97%

Supplier: AMBEED, INC
Warning

A250084-1G A250084-100MG A250084-250MG
103811-912EA 142 USD InStock
103811-912 77031-480 77031-482
1-Azaspiro[4.5]decane-2,8-dione 97%
1-Azaspiro[4.5]decane-2,8-dione
Precaución: for Research Use Only


Formula: C₉H₁₃NO₂
Boiling Pt: 401 °C (760 mm Hg)
Flash Pt: 190 °C
Storage Temperature: Refrigerator
MDL Number: MFCD14706827
CAS Number: 749861-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty