Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-3-methylpiperidin-4-one 97%

Supplier: AMBEED, INC
Danger

A205260-1G A205260-5G A205260-25G A205260-10G
103817-620EA 9 USD InStock
103817-620 103848-286 77086-494 77192-464
1-Benzyl-3-methylpiperidin-4-one 97%
1-Benzyl-3-methylpiperidin-4-one
Precaución: for Research Use Only


Formula: C₁₃H₁₇NO
Storage Temperature: Refrigerator
MDL Number: MFCD00044806
CAS Number: 34737-89-8
UN: 2735
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty