Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzyl-4-bromo-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A121957-1G A121957-100G A121957-10G A121957-25G A121957-1G A121957-5G
103800-158EA 11 USD InStock
103800-158 76995-878 76995-880 76995-882 77297-850 77312-290
1-Benzyl-4-bromo-1H-pyrazole 97%
1-Benzyl-4-bromo-1H-pyrazole
Precaución: for Research Use Only


Formula: C₁₀H₉BrN₂
Boiling Pt: 344.4 °C (760 mm Hg)
Melting Pt: 50…54 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02179566
CAS Number: 50877-41-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty