Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(4-formylphenyl)piperazine 98%

Supplier: AMBEED, INC
Warning

A119861-1G A119861-10G A119861-250MG A119861-25G A119861-1G
103799-848EA 6 USD InStock
103799-848 77026-316 77026-318 77026-320 77311-956
1-Boc-4-(4-formylphenyl)piperazine 98%
1-Boc-4-(4-formylphenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₆H₂₂N₂O₃
MW: 290.3600 g/mol
Boiling Pt: 441.4 °C (760 mm Hg)
Melting Pt: 118…121 °C
Flash Pt: 220.8 °C
Storage Temperature: Ambient
MDL Number: MFCD05864663
CAS Number: 197638-83-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty