Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(hydroxymethyl)-4-methylpiperidine 95%

Supplier: AMBEED, INC
Warning

A123210-1G A123210-250MG A123210-5G
103800-364EA 20 USD InStock
103800-364 77189-740 77312-482
1-Boc-4-(hydroxymethyl)-4-methylpiperidine 95%
1-Boc-4-(hydroxymethyl)-4-methylpiperidine
Precaución: for Research Use Only


Formula: C₁₂H₂₃NO₃
MW: 229.3200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08062516
CAS Number: 236406-21-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty