Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-4-(tosyloxy)piperidine 98%

Supplier: AMBEED, INC
Warning

A131837-1G A131837-10G A131837-250MG A131837-1G A131837-5G A131837-25G
103801-644EA 8 USD InStock
103801-644 77109-300 77109-302 77298-512 77298-514 77314-174
1-Boc-4-(tosyloxy)piperidine 98%
1-Boc-4-(tosyloxy)piperidine
Precaución: for Research Use Only


Formula: C₁₇H₂₅NO₅S
MW: 355.4500 g/mol
Boiling Pt: 475.3 °C (760 mm Hg)
Melting Pt: 100…104 °C
Flash Pt: 241.2 °C
Storage Temperature: Ambient
MDL Number: MFCD06796535
CAS Number: 118811-07-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty