Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-azetidine-3-ylmethanol 98%

Supplier: AMBEED, INC
Warning

A172912-1G A172912-5G A172912-100G A172912-10G A172912-25G A172912-5G A172912-1G
103806-762EA 13 USD InStock
103806-762 103845-054 77011-658 77011-660 77082-270 77082-272 77301-234
1-Boc-azetidine-3-ylmethanol 98%
1-Boc-azetidine-3-ylmethanol
Precaución: for Research Use Only


Formula: C₉H₁₇NO₃
Boiling Pt: 270.3 °C (760 mm Hg)
Melting Pt: 56…60 °C
Flash Pt: 117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06656141
CAS Number: 142253-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty