Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Boc-piperidine 98%

Supplier: AMBEED, INC
Danger

A199786-1G A199786-5G A199786-1G A199786-25G
103817-134EA 9 USD InStock
103817-134 103847-760 77302-628 77302-630
1-Boc-piperidine 98%
1-Boc-piperidine
Precaución: for Research Use Only


Formula: C₁₀H₁₉NO₂
Boiling Pt: 60…65 °C (760 mm Hg)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD02093935
CAS Number: 75844-69-8
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty